Learn More
Gelatin, Granular, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Gelatin derived from porcine skin is a powder created from the acidic digestion of collagen and is also known as gelatin type A
• It comprises heterogeneous mixture of water-soluble proteins of high average molecular masses, including glycine, proline and hydroxyproline
Applications
• Gelatin has been used in in vitro coating cell culture to improve attachment of cell lines • It is added to PCR reactions in order to stabilize DNA Taq polymerase
• It can be used as a blocking reagent in several biological techniques including western blotting, ELISA, and immunochemistry
• It can be applied in microencapsulation assays to delivery vehicle for release of active biomolecules since it is a biocompatible polymer
• It is used as a component of media for species differentiation in bacteriology and in light microscope autoradiography
• It has been used in the generation of scaffolds for tissue engineering assays

Specifications
Specifications
Color | Beige to Yellow |
pH | 3.8 to 7.6 (1% soln., about 55°C) (PHEUR) |
Loss on Drying | 13% max. (105°C) |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Quantity | 2.5 kg |
Merck Index | 15, 4415 |
Grade | Analysis |
Physical Form | Crystalline Powder |
Chemical Name or Material | Gelatin |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464467 | Certificate of Analysis | 11/07/2024 | 410875000,410870025 |
A0460301 | Certificate of Analysis | 22/02/2024 | 410875000,410870025 |
A0453401 | Certificate of Analysis | 25/08/2023 | 410875000,410870025 |
A0448465 | Certificate of Analysis | 30/12/2022 | 410875000,410870025 |
A0438935 | Certificate of Analysis | 09/02/2022 | 410875000,410870025 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.