missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GelCode™ Blue Safe Protein Stain

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and membranes.

160.00 € - 325.00 €

Specifications

Content And Storage Store at room temperature.
Detection Location In-Blot Detection, In-Gel Detection
Color Blue
Detection Method Colorimetric
Label or Dye Coomassie
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10658884
View Documents
Thermo Scientific™
24594
1 L
160.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10763505
View Documents
Thermo Scientific™
24596
3.5 L
325.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and membranes. This protein-specific stain allows bands to be viewed directly on a gel or membrane during staining and is compatible with mass spectrometry applications. GelCode Blue Safe Protein Stain is non-corrosive, non-flammable, and is classified as non-hazardous under the U.S. Department of Transportation (DOT) shipping requirements.

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and is formulated to avoid hazardous shipping.

Features include:
Sensitive—detect down to 9 ng of protein per band in 15 minutes
Safe—non-corrosive to skin, non-flammable, and safe to ship and store until activated
Minimal additional reagents—water washes only; no acid-fixative or methanol destain solutions required
Compatible—suitable for staining 1D and 2D polyacrylamide gels and nitrocellulose and PVDF membranes (multiple protocols provided in the instructions)
Multiple applications—qualitative visualization, quantitative densitometry, and downstream excision and analysis by mass spectrometry
Easy to use—add and dissolve activator crystals before first use, then simply add reagent to stain gels
Stable—store non-activated or activated stain at room temperature for up to one year

Because the supplied activator crystals are packaged and shipped dry in a separate bottle, GelCode Blue Safe Protein Stain is classified as nonhazardous under the U.S. Department of Transportation (DOT) shipping requirements. For proper protein staining, the activator crystals must be dissolved in the staining reagent before use. Add the entire contents of the activator crystals bottle into the GelCode Blue Safe Protein Stain bottle. Dissolve the crystals by mixing end-over-end for one minute, then allow it to remain at room temperature for five minutes before use. Store the activated stain at room temperature.

Specifications

Specifications

Store at room temperature.
Blue
Coomassie
GelCode™
In-Blot Detection, In-Gel Detection
Colorimetric
Safe Protein Stain
Protein
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GelCode™ Blue Safe Protein Stain

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.