Learn More
Thermo Scientific™ GelCode™ Blue Stain Reagent
GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.
Brand: Thermo Scientific™ 24590
99.40 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection. This unique reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process.
GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.Features include:
• No methanol-acetic acid destaining required
• Fast, one-step, one-hour staining, ready-to-use stain reagent; band development visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
• Water wash increases staining sensitivity; optional step provides crystal-clear gel background and makes weakly stained bands more visible
GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.
Specifications
Store at 4°C. | |
In-Blot Detection, In-Gel Detection | |
Coomassie | |
GelCode™ | |
Protein |
GelCode™ Blue Stain Reagent, 500 mL | |
Colorimetric | |
Protein Gel Stain Reagent | |
500 mL |
Safety and Handling
- GelCode Blue Stain Reagent
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H371-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol, Phosphoric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.