Learn More
Tocris Bioscience™ Geldanamycin
Selective Hsp90 inhibitor
Brand:  Tocris Bioscience™ 1368/1
392.20 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18065182
  • 431.00 € / 1mg
Explore more special offers
Description
Description
Selectively inhibits heat shock protein 90 (Hsp90). Binds to the ATP site of Hsp90 (Kd = 1.2 μM) and inhibits its chaperone activity. Consequently inhibits activities of oncogenic kinases (e.g. src, Raf), p53 and steroid receptors. Demonstrates antiproliferative effect on breast cancer stem-like cells.
Specifications
Specifications
1 mg
>98%
C29H40N2O9
Soluble to 10 mM in DMSO
Desiccate at -20°C
30562-34-6
560.64g/mol