missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11644-2-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
Gelsolin | |
Polyclonal | |
Unconjugated | |
GSN | |
Actin depolymerizing factor, ADF, AGEL, Brevin, Gelsolin, GSN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227753, 2934, 296654 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P06396, P13020, Q68FP1 | |
Gsn | |
Gelsolin Fusion Protein Ag2239 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |