missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer

Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Brand:  Applied Biosystems™ N8080006

 View more versions of this product

Product Code. 10219244

  • 52.50 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applied Biosystems™ GeneAmp™ 10X PCR Buffer I is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes

GeneAmp™ 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq™ DNA Polymerase.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• GeneAmp 10X PCR Buffer I (with 15 mM MgCl2), 1.5 mL

Store at -15°C to -25°C.
PCR Buffer
Dry Ice
10X
Kit
AmpliTaq DNA Polymerase
1.5 mL
Standard PCR
10X
1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.