missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GeneArt™ Linear pUC19L Vector for Seamless Cloning
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
Description
The GeneArt™ Linear pUC19L Vector is a ready-to-use, linear version of the popular cloning vector pUC19. The pUC19L plasmid (2.7 Kbp), is specifically designed as a cloning and DNA assembly vector for the GeneArt™ Seamless Cloning and Assembly Kit (cat # A13288). The vector is manufactured by digesting supercoiled pUC19 plasmid with the restriction enzymes PstI and KpnI, and tested for cloning of up to 4 DNA fragments.
For Research Use Only. Not for use in diagnostic procedures.
- Some of the key features and elements of the pUC19L vector include:
- lacZα gene allows easy blue⁄white colony screening for selection of recombinants
- pUC origin of replication (high copy vector)
- Ampicillin resistance for selection in E. coli
- 6 convenient and validated restriction sites flanking the cloning region
- Tested for functionality by assembly with 4 DNA fragments using the GeneArt™ Seamless Cloning and Assembly Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Content And Storage | Store the product at -80°C. The vector is guaranteed stable for 6 months when properly stored. |
| Description | Reporter: Beta-Gal (lacZ) |
| For Use With (Application) | PCR cloning, seamless cloning and genetic assembly |
| Includes | The product contains 5 vials of linear pUC19L vector at a concentration of 50ng⁄ml. Each vial contains enough plasmid for 4 cloning reactions using the GeneArt™ Seamless Cloning and Assembly Kit (A13288 - must be purchased separately). |
| No. of Reactions | 20 |
| Product Type | GeneArt™ Linear pUC19L Vector for Seamless Cloning |
| Quantity | 20 Reactions |
| Sample Volume | 13 kb total (vector plus all inserts), Up to 4 Fragments |
| Vector | Seamless Cloning Vector, Linear |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction