Learn More
Thermo Scientific™ GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 Cells
GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells contain the human Adenosine Receptor A2A (ADORA2A) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1530
Product Code. 10626414
  • 17190.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells contain the human Adenosine Receptor A2A (ADORA2A) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of NECA. In addition, GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
ADORA2A has been shown to have many roles throughout the body. ADORA2A is involved in pain and inflammation regulation, inhibition of platelet aggregation, blood pressure regulation and contributes to ischemic brain damage. Selective antagonists of the ADORA2A pathway are being tested as adjuvants to dopaminergic drugs to treat Parkinson's disease and schizophrenia.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000675
Dividing Cells
Fluorescence
ADORA2A
Single Cell Clone Origin
2 x 106 Cells
ADORA2A
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells
• GeneBLAzer™ ADORA2A-CRE-bla CHO-K1
• Certificate of Analysis
The GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
ADORA2A-CRE-bla CHO-K1 cells, ADORA2A-CRE-bla CHO-K1, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
Dividing Cells
GeneBLAzer™