Learn More
Thermo Scientific™ GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F Cells
The GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells contain the human Amylin Receptor 2 (CALCRL:RAMP3), stably integrated into the CellSensor™ CRE-bla Freestyle™ 293F cell line.
Brand:  Thermo Scientific™ K1439
Product Code. 10552194
  • 16120.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells contain the human Amylin Receptor 2 (CALCRL:RAMP3), (Accession # NM_005795.3 NM_005856.1) stably integrated into the CellSensor™ CRE-bla Freestyle™ 293F cell line. CellSensor™ CRE-bla Freestyle™ 293F cells (Cat. No. K1130) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).
CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells are functionally validated for Z'-factor and EC50 concentrations of Adrenomedullin(1-52). In addition, CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_005855.2
Dividing Cells
GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F Cells, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, FreeStyle™ 293-F Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F Cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F cells, GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F Assay Protocol, Certificate of Analysis
Freestyle™, GeneBLAzer™
BLA (Beta-Lactamase)
CALCRL:RAMP3
FreeStyle 293-F Cells
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F cells
• GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
CALCRL:RAMP3
Single Cell Clone Origin
8 x 106 Cells
FreeStyle™ 293-F
Cell Line
GeneBLAzer™