Learn More
Thermo Scientific™ GeneBLAzer™ CCKAR-NFAT-bla HEK 293T Cells
The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1548
Product Code. 10781814
  • 15770.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.
The CCKAR-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, CCKAR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000730.2
Dividing Cells
Fluorescence
CCKAR
Single Cell Clone Origin
2 x 106 Cells
CCKAR
293T (HEK)
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ CCKAR-NFAT-bla HEK 293T cells
• GeneBLAzer™ CCKAR-NFAT-bla HEK 293T Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ CCKAR-NFAT-bla HEK 293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
GPCRs
CCKAR-NFAT-bla HEK 293T cells, CCKAR-NFAT-bla HEK 293T Assay Protocol, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
Dividing Cells
GeneBLAzer™