missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ EDG7 HEK 293T DA Assay Kit

GeneBLAzer™ EDG7 HEK 293T DA (Division Arrested) cells and EDG7-NFAT-bla HEK 293T cells contain the human EDG7 receptor stably integrated in the CellSensor™ NFAT-bla HEK 293T cell line.

Brand:  Thermo Scientific™ K1315

Product Code. 10267173

  • 996.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This cell line is available as dividing cells by requesting a quote for Cat. No. K1251 from dividingcells@invitrogen.com

GeneBLAzer™ EDG7 HEK 293T DA (Division Arrested) cells and EDG7-NFAT-bla HEK 293T cells contain the human EDG7 receptor stably integrated in the CellSensor™ NFAT-bla HEK 293T cell line. CellSensor™ NFAT-bla HEK 293T cells (Cat. No. K1179) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both EDG7 HEK 293T DA cells and EDG7-NFAT-bla HEK 293T cells are functionally validated for Z'-factor and EC50 concentrations of Lysophosphatidic acid (LPA); (Figure 1). In addition, EDG7-NFAT-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

LPA receptors were originally classified in the Endothelial Differentiation Gene (EDG) family of receptors. The EDG family was later divided into two distinct groups of receptors based on their ligand specificity for either Lysophosphatidic acid (LPA) or Sphingosine-1-Phosphate (S1P). There are three high affinity LPA receptors LPA1, LPA2, and LPA3 (formally EDG2, EDG4, and EDG7), and five high affinity S1P receptors S1P1, S1P2, S1P3, S1P4 and S1P5 (formally EDG1, EDG5, EDG3, EDG6, and EDG8). LPA receptors are differentially linked via Gi and Gq to multiple effector systems and diverse biological functions. Recent studies revealed that targeted deletion of the EDG7/LPA3 receptor results in delayed implantation and altered embryo spacing, and significantly reduced litter size in mice. These findings indicate that EDG7 plays a crucial role in implantation and mammalian reproduction.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_012152.1
Division-Arrested Cells
Gene ID: 23566
GPCRs
EDG7
GeneBLAzer™
BLA (Beta-Lactamase)
Division-Arrested Cells Assay Kit
GeneBLAzer™
Cell-based beta lactamase reporter gene

GeneBLAzer™ EDG7 HEK 293T DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• EDG7 HEK 293T DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ EDG7 HEK 293T DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDG7 HEK 293T DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ EDG7-NFAT-bla HEK 293T cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDG7-NFAT-bla HEK 293T cells
• 1 protocol
• Certificate of Analysis

Storage
GeneBLAzer™ EDG7 HEK 293T DA cells and EDG7-NFAT-bla HEK 293T cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.

Fluorescence
23566
Single Cell Clone Origin
1 Plate
EDG7
293T (HEK)
GPCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.