missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ ERβ-UAS-bla GripTite™ Cells

ER beta-UAS-bla GripTite™ 293 cells contain the ligand-binding domain (LBD) of the human Estrogen receptor beta (ER beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla GripTite™ 293 cell line.

Brand:  Thermo Scientific™ K1689B

Product Code. 10145793

  • 10380.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ER beta-UAS-bla GripTite™ 293 cells contain the ligand-binding domain (LBD) of the human Estrogen receptor beta (ER beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla GripTite™ 293 cell line. GeneBLAzer™UAS-bla GripTite™ 293 cells stably express a betalactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-ER beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. ER beta-UAS-bla GripTite™293 cells are functionally validated for Z' and EC50 concentrations of 17-beta Estradiol. In addition, ER beta-UAS-bla GripTite™293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dividing Cells
Fluorescence, Fluorescent
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
GeneBLAzer™ ER beta -UAS-bla GripTite™ cells Includes: ER beta-UAS-bla GripTite™ cells, Protocol, Certificate of Analysis
GeneBLAzer™
GripTite Cells
BLA (Beta-Lactamase)
Cell Line
ER beta-UAS-bla GriptTite™ cells (K1091)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen), Liquid nitrogen, room temperature
GeneBLAzer™ ERβ-UAS-bla GripTite™ Cells, 2 x 106 Cells, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Nuclear Receptors
2 x 106 Cells
2 x 106 Cells
GripTite™ 293
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.