missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ GR DA Assay Kit

GeneBLAzer™GR DA (Division Arrested) cells and GR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Gluocorticoid receptor (GR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

Brand:  Thermo Scientific™ K1391

Product Code. 10655685

  • 1516.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

GeneBLAzer™GR DA (Division Arrested) cells and GR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Gluocorticoid receptor (GR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-blaHEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-GR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have noapparent toxicity or change in cellular signal transduction. Both GR DA cells and GR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of Dexamethasone. In addition, GR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetrac, T4
Cell-based beta lactamase reporter gene
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• GR HEK 293T DA cells (K1391A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Fluorescence
2908
GR HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg
GeneBLAzer™
BLA (Beta-Lactamase)
NR3C1, GR
293T (HEK)
Nuclear Receptors
1-850
Division-Arrested Cells
GeneBLAzer™ GR DA Assay Kit, 1 Plate, ∽9,600,000 cells/vial Quantity of Cells, 293T (HEK) Cell Line, 1-850 Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays
GR
T3 Hormone
1 Plate
293T (HEK)
Division-Arrested Cells Assay Kit
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.