Learn More
Thermo Scientific™ GeneBLAzer™ HTR7-CRE-bla CHO-K1 Cells
GeneBLAzer™ HTR7 CHO-K1 DA (Division Arrested) cells and HTR7-CRE-bla CHO-K1 cells contain the human 5-hydroxytryptamine (serotonin) receptor 7 stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1704
Product Code. 10658714
  • 16660.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
GeneBLAzer™ HTR7 CHO-K1 DA (Division Arrested) cells and HTR7-CRE-bla CHO-K1 cells contain the human 5-hydroxytryptamine (serotonin) receptor 7 (Accession No. NM_019860.2) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase (bla) reporter gene under control of the Cyclic AMP Response Element (CRE).
GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of 5-hydroxytryptamine (5-HT). In addition, GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_019860.2
Dividing Cells
GeneBLAzer™ HTR7-CRE-bla CHO-K1 Cells, 2 x 106 Cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
FRET
GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells, GeneBLAzer™ HTR7-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
HTR7
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells
• GeneBLAzer™ HTR7-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
HTR7
Single Cell Clone Origin
2 x 106 Cells
CHO-K1
CHO-K1 Cells
GeneBLAzer™