Learn More
Invitrogen™ GeneBLAzer™ In Vivo Detection Kit
GeneBLAzer™ In Vivo Detection Kit
Brand:  Invitrogen™ 12578134
Product Code. 10432862
  • 453.00 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The GeneBLAzer™ In Vivo Detection Kit measures β-lactamase reporter activity in live mammalian cells, using the β-lactamase substrate CCF2-AM. The GeneBLAzer™ Detection System provides the following advantages:
  • Suitable for use as a sensitive reporter of gene expression in living mammalian cells using fluorescence microscopy.
  • Provides a ratiometric readout to minimize differences due to variability in cell number, substrate concentration, fluorescence intensity, and emission sensitivity
  • Uses a non-toxic substrate, allowing continued cell culturing after quantitative analysis.
  • Compatible with a wide variety of applications, including microplate-based transcriptional assays and flow cytometry.
  • Provides a flexible and simple assay development platform for gene expression in mammalian cells.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    GeneBLAzer™
    Fluorescence
    375 Reactions
    Reporter Assays
    Beta-Lactamase Substrate
    In vivo detection kits contain 50 μg of CCF2-AM substrate (enough for 375 96-well reactions). Substrate should be stored at -20°C, dessicated, and protected from light. All reagents are guaranteed stable 6 months when properly stored.
    Detection Kit
    Kit
    BLA (Beta-Lactamase)