Learn More
Thermo Scientific™ GeneBLAzer™ LXR beta DA Assay Kit
GeneBLAzer™LXR beta DA (Division Arrested) cells and LXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-beta (LXR beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.
Brand: Thermo Scientific™ K1415
Description
GeneBLAzer™LXR beta DA (Division Arrested) cells and LXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-beta (LXR beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-LXR beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both LXR beta DA cells and LXR beta-UAS-bla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of TO901317. In addition, LXR beta-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Specifications
DHT | |
Cell-based beta lactamase reporter gene | |
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate: • LXR beta HEK 293T DA cells (K1415A) • LiveBLAzer™-FRET B/G Loading Kit, 70 μg Shipping: Dry Ice Storage: -80°C (liquid nitrogen) |
|
Fluorescence | |
7376 | |
LXR beta HEK 293T DA cells (K1415A) LiveBLAzer™-FRET B/G loading kit | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
LXR beta, NR1H2, LXRB | |
293T (HEK) | |
Nuclear Receptors |
Cyp Acetate, Spirolactone, OHF= | |
Division-Arrested Cells | |
GeneBLAzer™ LXR beta DA DA Assay Kit, 1 Plate, ∽4, 800,000 No. of Cell, Liquid, Division-Arrested Cells State, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Target, BLA (Beta-Lactamase) Reporter, Epithelial Cell Phenotype, Adherent Growth Properties | |
Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays | |
LXRβ | |
R1881 | |
1 plate | |
293T (HEK) | |
Division-Arrested Cells Assay Kit | |
GeneBLAzer™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.