Learn More
Thermo Scientific™ GeneBLAzer™ MCHR2-NFAT-bla CHO-K1 Cells
GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells contain the human melanin-concentrating hormone receptor 2 (GPR145) stably integrated into the CellSensor NFAT-bla CHO-K1 parental cell line.
Brand:  Thermo Scientific™ K1852
Product Code. 10216894
  • 15150.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells contain the human melanin-concentrating hormone receptor 2 (GPR145) stably integrated into the CellSensor NFAT-bla CHO-K1 parental cell line. CellSensor NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of a NFAT response element stably integrated into the cells. GeneBLAzer MCHR2-NFAT-bla CHO-K1 cells have been functionally validated for Z'-Factor and EC50 concentrations of MCH 6-17.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_032503.2
Dividing Cells
Fluorescence
G-Protein Coupled Receptor (GPCR)
GeneBLAzer™
End Point
Dividing Cells
GeneBLAzer™
Cell-based beta lactamase reporter gene
GeneBLAzer™ MCHR2-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 106 cells/vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
FRET
Single Cell Clone Origin
1 Vial
GPR145, MCHR2
CHO-K1
GPCR