missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ MR-UAS-bla HEK 293T Cells

MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR) fused to the NA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

Brand:  Thermo Scientific™ K1696

Product Code. 10339243

  • 10680.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR) fused to the NA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-MR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384- well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. MR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of aldosterone. In addition, MR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and substrate loading time. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

AM 580, Fenretinide, 9-cis- retinoic acid, Ch 55, TTNPB
Cell-based beta lactamase reporter gene
MR-UAS-bla HEK 293T cells (K1696)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen), Liquid Nitrogen
GeneBLAzer™ MR-UAS-bla HEK 293T Cells, 2 x 106 Cells, LE 135 Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Nuclear Receptors
2 x 106 Cells
GeneBLAzer™
HEK 293T Cells
BLA (Beta-Lactamase)
Cell Line
LE 135
Dividing Cells
Fluorescence, Fluorescent
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
GeneBLAzer™ MR-UAS-bla HEK 293T cells Includes: MR-UAS-bla HEK 293T cells, Protocol, Certificate of Analysis
All-trans-retinoic Acid
2 x 106 Cells
293T (HEK)
NR3C2, MR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.