Learn More
Thermo Scientific™ GeneBLAzer™ RARγ-UAS-bla HEK 293T Cells
The GeneBLAzer™ RAR gamma-UAS-bla HEK 293T cells contain the ligand binding domain (LBD) of the human retinoic acid receptor gamma (RAR gamma) fused to the DNA-binding domain of GAL4, stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line.
Brand:  Thermo Scientific™ K1763
Product Code. 10043963
  • 11040.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The GeneBLAzer™ RAR gamma-UAS-bla HEK 293T cells contain the ligand binding domain (LBD) of the human retinoic acid receptor gamma (RAR gamma) fused to the DNA-binding domain of GAL4, stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells (Cat. No. K1104) stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RAR gamma (LBD) fusion protein, it binds to the UAS, inducing transcription of beta-lactamase. GeneBLAzer™ RAR gamma-UAS-bla HEK 293T cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. The Z' and EC50 concentrations of all-trans-retinoic acid and other stimuli have been determined. Additional testing data are available upon request.
RAR gamma is a member of the retinoic acid receptor family of nuclear receptors. Retinoids, vitamin A and its natural and synthetic analogues, are a very important group of hormones that regulate a wide variety of biological functions including embryogenesis, cell growth, and cell differentiation. Retinoids are beneficial in a wide variety of clinical cases including dermatology, oncology, diabetes, and diseases associated with HPV. RAR gamma is the predominant RAR subtype present in the skin and it constitutes more than 90% of the total skin or keratinocyte RAR repertoire. Skin irritation is a classical toxicity associated with RAR pan-agonists, like RA or TTNPB, and is believed to be a manifestation of RAR gamma activation. Furthermore, it has been shown that tumor-specific apoptosis can be driven by RAR gamma selective agonists, for pancreatic cancer cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-hydroxytamoxifen, Chlordane, Toxaphene
Dividing Cells
Fluorescence, Fluorescent
Nuclear Receptors
XCT790
2 x 106 Cells
BLA (Beta-Lactamase)
Dividing Cells
Cell-based beta lactamase reporter gene
GeneBLAzer™ Nuclear Receptor Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use., Liquid Nitrogen
Nuclear Receptors
2 x 106 Cells
GeneBLAzer™
HEK 293T Cells
RARG, RAR gamma, NR1B3