missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ RXR alpha DA Assay Kit

GeneBLAzer™ RXR alpha DA (Division Arrested) cells and RXR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoid X receptor-alpha (RXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UASbla HEK 293T cell line.

Brand:  Thermo Scientific™ K1411

Product Code. 10358624

  • 1574.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

GeneBLAzer™ RXR alpha DA (Division Arrested) cells and RXR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoid X receptor-alpha (RXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UASbla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a betalactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RXR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of alpha-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both RXR alpha DA cells and RXR alpha-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of 9-cis-retinoic acid. In addition, RXR alpha-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dexamethasone, Progesterone, Cortisol
Cell-based beta lactamase reporter gene
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• RXR alpha HEK 293T DA cells (K1411A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Fluorescence
RXRα
Aldosterone
1 plate
293T (HEK)
Division-Arrested Cells Assay Kit
GeneBLAzer™
Spironolactone
Division-Arrested Cells
GeneBLAzer™ RXR alpha DA Assay Kit, 1 Plate, ∽4,800,000 cells/vial at 4,800,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, Spironolactone Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays
RXR alpha HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg
GeneBLAzer™
BLA (Beta-Lactamase)
RXRA, RXR alpha, NR2B1
293T (HEK)
Nuclear Receptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.