missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ VPAC2-CRE-bla CHO-K1 Cells

GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells contain the human Vasoactive intestinal peptide/pituitary adenylate cyclase activating polypeptide receptor 2 (VPAC2) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

Brand:  Thermo Scientific™ K1706

Product Code. 10093913

  • 16120.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CellSensor™ CRE-bla CHO-K1 cells (Cat. no. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of VIP. In addition, GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

VPAC2 has been implicated in a wide range of autoimmune and inflammation disorders, including Chronic obstructive pulmonary disease, Chronic Fatigue Syndrome. VPAC2 may also play a role in Type-2 diabetes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_003382.3
Dividing Cells
Fluorescence
VPAC2
GeneBLAzer™
BLA (Beta-Lactamase)
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
Contents:
K1706 GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells
• GeneBLAzer™ VPAC2-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ VPAC2-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
Single Cell Clone Origin
2 x 106 Cells
VPAC2
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.