missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ GeneClean Kit

GeneClean™ Kit, Solution or Gels

Brand:  MP Biomedicals™ 111001200

272.18 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.



Product Code. 15233667

  • 322.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The GENECLEAN™ Kit includes Glassmilk™ slurry that allows flexibility of reaction size and volume.
  • The GENECLEAN™ kit simplifies the process of purifying DNA into three easy steps: Bind,Wash and Elute.
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
  • Isolate DNA from TAE buffered agarose gels and solutions
  • The GENECLEAN™ Kit is used to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

New Wash/Ethanol Concentration must be ∽ 50%
Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away fromgenomic DNA.
Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Solution or Gels
Store at Room Temperature (15-30°C). NaI solution must be kept away from light and open air sources to prevent oxidation. NaI is packaged under nitrogengas to further reduce chance of oxidation. Shelf life may be extended by storing at 4°C.
200 Preps
Kits and Assays
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneClean Kit

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium iodide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers