missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET RNA Purification Kit

Thermo Scientific GeneJET RNA Purification Kit is a simple and efficient system for purification of total RNA from mammalian cultured cells, tissue, human blood cells, bacteria, and yeast.

Brand:  Thermo Scientific™ K0732


 View more versions of this product

Product Code. 10171080

  • 1266.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific GeneJET RNA Purification Kit is a simple and efficient system for purification of total RNA from mammalian cultured cells, tissue, human blood cells, bacteria, and yeast.

The kit utilizes a silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious cesium chloride gradients, alcohol precipitation, or toxic phenol-chloroform extractions.

RNA molecules longer than 200 nucleotides can be isolated with the GeneJET RNA Purification Kit in 15 minutes after the lysis step. The high-quality purified RNA can be used in a wide range of downstream applications, including RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analyses.

Highlights
Universal—can be used for both cell and tissue samples from a wide range of sources: mammalian blood, mammalian cell cultures, mammalian tissues, insect, yeast, and bacteria
Efficient—high yields of total RNA.
Fast—15 minutes after lysis step
Pure—an A260/280 ratio > 1.9
Convenient—spin columns are capped and assembled with collection tubes

Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Includes
Proteinase K Lysis Buffer, Wash Buffer 1 (concentrated), Wash Buffer 2 (concentrated), Nuclease-free Water, GeneJET RNA Purification Columns (pre-assembled with Collection Tubes), Collection Tubes (2 mL), Collection Tubes (1.5 mL), Detailed Protocol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 5 x 600 μL Proteinase K; store at room temperature until opened, -20°C after opening
• 200 mL Lysis Buffer; room temperature
• 200 mL Wash Buffer 1 (concentrated); room temperature
• 100 mL Wash Buffer 2 (concentrated); room temperature
• 125 mL Nuclease-free Water; room temperature
• 250 GeneJET RNA Spin Columns with Collection Tubes; room temperature
• 250 Collection Tubes (2 mL); room temperature
• 250 Collection Tubes (1.5 mL); room temperature
15 min. (After sample homogenization)
Bacteria, Blood, Cells, Tissue, Yeast, Insects, Enzymatic Reactions
Not High-throughput Compatible (Manual)
250 Preps
Room Temperature
Up to 160 μg
Silica Spin Column
50–100 μL
Total RNA
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Mini
Bacteria: ≤109 cells
Blood: 0.5 mL
Cells: ≤107
Tissue and Insects: 30 mg
Yeast: ≤4 x 108 cells
Enzymatic reactions: ≤100 μL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneJET RNA Purification Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, Guanidine hydrochloride, Proteinase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.