Learn More
Corning™ General Assay Microplate Lids
GREENER_CHOICE
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand:  Corning™ 3098
225.07 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 10157051
  • 286.60 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Polystyrene
    100/Cs.
    Universal Lid
    No
    Sterile
    25/Pk.
    All Microplates
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Information
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ General Assay Microplate Lids > Universal Lid without Corner Notch; 100/Cs