missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 120 - General Purpose Digital Heating Circulator Stirrer with Integrated Pump for External Applications

This general purpose digital heated circulator provides great temperature stability and control for demanding applications requiring accurate temperature control. Operates over the range of ambient +5°C to 120°C or down to -15°C with optional accessory cooling.

Brand:  Fisherbrand™ 17225566

Product Code. 17225566

  • 1105.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Temperature ranges from ambient 5°C to 120°C (using baths larger than 5 L), to ambient 15°C to 120°C (with 5 L baths)
  • Cooling/heating capacities can be reached with a cooling unit
  • Simple, intuitive user interface with dial and two function buttons
  • Clear, bright 4-digit LED display
  • Integrated pump for external application
  • Visual alarm and low liquid detection
  • 2-point user calibration
  • 3 temperature presets

Applications:

  • Clinical, Microbiology and Pathology labs for media tempering, thawing, and incubating samples
  • University research for temperature control of spectrophotometers, refractometers and jacketed vessels
  • Industrial labs for temperature probe calibration, water analysis, QC testing product, petrochemical testing, material testing, and milk sample testing
Specifications

Specifications

Digital
-15°C (with optional immersion cooler)
-20°C to +120°C
4-digit LED
210 mbar
5.1 lb.
230 V
50/60 Hz
Air-cooled
16 L/min.
Heating Circulator
Yes
±0.05°C
2.3 kg
1300 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.