missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 100 - General Purpose Digital Heating Circulator Stirrer

This general purpose digital heated circulator provides great temperature stability and control for demanding applications requiring accurate temperature control. Operates over the range of ambient +5°C to 100°C or to 0°C with optional accessory cooling.

Brand:  Fisherbrand™ 17265556

Product Code. 17265556

This item is not returnable. View return policy

Description

Description

  • Temperature ranges from ambient 5°C to 100°C (using baths larger than 5 L), to ambient 15°C to 100°C (with 5 L baths)
  • Cooling/heating capacities can be reached with a cooling unit
  • Simple, intuitive user interface with dial and two function buttons
  • Clear, bright 4-digit LED display
  • Visual alarm and low liquid detection
  • 2-point user calibration
  • 3 temperature presets

Applications:

  • Clinical, Microbiology and Pathology labs for media tempering, thawing, and incubating samples
  • University research for temperature control of spectrophotometers, refractometers and jacketed vessels
  • Industrial labs for temperature probe calibration, water analysis, QC testing product, petrochemical testing, material testing, and milk sample testing

Order Info

Stainless steel bath ordered separately.

Specifications

Specifications

13.2 gal.
Digital
0 to 13 L/min.
Heating Circulator
No
2.1 kg
1.4/1.5 kW
50 L
Air-cooled
0°C to 100°C
4-digit LED
4.6 lb.
120/230 V
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers