Learn More
3M™ General Purpose Masking Tape 201E
The 3M™ General Purpose Masking Tape 201E is a premium-quality masking tape used for short-term paint masking, fixing, bundling, holding, sealing, and other applications up to a temperature of 80°C for one hour.
89.00 € - 125.00 €
Specifications
Color Beige
Material Crepe Paper
Product Type Masking Tape
View More Specs
Products 2
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
17843256
View Documents
3M™
7100195888
50 m x 48 mm
125.00 €
Pack of 24
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17833256
View Documents
3M™
7100195952
50 m x 24 mm
89.00 €
Pack of 36
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
Description
Description
  • This tape is designed for general use in industrial applications with a slight need for temperature and water resistance such as short-term paint masking and sealing applications.
  • It provides a good instant adhesion and holding power that resists lifting or curling.
  • Its crepe paper backing offers conformability that will maintain integrity when formed around corners.
  • Its good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines due to its clean removability in one piece with no residue from most surfaces.
Specifications
Specifications
Beige
Crepe Paper
Masking Tape
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ General Purpose Masking Tape 201E