missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General-Purpose Narrow-Neck Bottles

Product Code. 11587433
Click to view available options
Capacity (Metric):
30 mL
125 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Use these general purpose bottles for liquid storage and sampling. Fisherbrand™ General-Purpose Narrow-Neck Bottles offer excellent chemical resistance. Bottles can be recycled.

  • Clear general purpose bottles
  • Polypropylene screw closure

Specifications

Capacity (Metric) 1000 mL
Color Translucent
For Use With (Application) For liquid storage and sampling
Material Polypropylene
Closure Included Yes
Mouth Narrow
Product Type Wash Bottle
Closure Material Polypropylene
Closure Size 28/R6
Diameter (Metric) 83 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'