Learn More
3M™ General Purpose Vinyl Tape 764
This vinyl tape is a cost-effective choice for a variety of hazard, safety marking, and color-coding applications when you’re not dealing with tough conditions or the need for a long-term clean removal. Comes individually wrapped and conveniently packaged.
Brand:  3M™ 7100260288
 View more versions of this product
Product Code. 17791689
  • 119.00 € / 12 rolls
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for marking applications in light traffic areas, walls and pipes
  • Use for taping in all non-critical applications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Protects parts, equipment and products from abrasions
  • Bundle and temporarily mark parts, machines, and more
  • Easy-to-apply tape stretches around corners for a reliable, tight fit
  • Fast tack and high holding strength
  • Available in wide variety of colors
  • Can be applied by hand or with floor applicator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
33 m x 50 mm
0.12 mm
Blue
12 Rolls
Rubber-based Adhesive
Vinyl