missing translation for 'onlineSavingsMsg'
Learn More

General Purpose Viscosity Standards, Paragon Scientific™

Certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation.

84.00 € - 126.00 €

Specifications

Quantity 500 mL
View More Specs

Products 26
Product Code Brand Viscosity Price Quantity & Availability  
Product Code Brand Viscosity Price Quantity & Availability  
16010272
View Documents
Paragon Scientific
N.4
0.2950 mPa.s (25°C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15427840
View Documents
Paragon Scientific
N.8
0.5204 mPa.s (25°C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15417840
View Documents
Paragon Scientific
N1.0
0.9525 mPa.s (25 °C)
87.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11994285
View Documents
Paragon Scientific
N2
2.144 mPa.s (25°C)
87.25 €
500mL
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
11999748
View Documents
Paragon Scientific
S3
3.291 mPa.s (25°C)
84.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11989748
View Documents
Paragon Scientific
S6
7.442 mPa.s (25°C)
87.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15341214
View Documents
Paragon Scientific
N7.5
8.559 mPa.s (25°C)
87.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
12166540
View Documents
Paragon Scientific
N1.0
14.42 mPa.s (25°C)
92.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11919758
View Documents
Paragon Scientific
S20
29.04 mPa.s (25°C)
84.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15524156
View Documents
Paragon Scientific
N26
48.59 mPa.s (25 °C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16020272
View Documents
Paragon Scientific
N35
55.36 mPa.s (25 °C)
92.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15781718
View Documents
Paragon Scientific
N44
71.11 mPa.s (25 °C)
91.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11969758
View Documents
Paragon Scientific
S60
100.7 mPa.s (25°C)
126.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15791718
View Documents
Paragon Scientific
N7.5
124.9 mPa.s (25 °C)
97.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16070260
View Documents
Paragon Scientific
N100
202.6 mPa.s (25 °C)
107.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16050370
View Documents
Paragon Scientific
S200
392 mPa.s (25°C)
125.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15555806
View Documents
Paragon Scientific
N250
488.1 mPa.s (25 °C)
87.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16000280
View Documents
Paragon Scientific
N350
717.4 mPa.s (25°C)
84.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15534156
View Documents
Paragon Scientific
N415
796.5 mPa.s (25 °C)
87.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16030380
View Documents
Paragon Scientific
S600
1273 mPa.s (25°C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16080260
View Documents
Paragon Scientific
N1000
2610 mPa.s (25°C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15592600
View Documents
Paragon Scientific
N1400
3481 mPa.s (25 °C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16020280
View Documents
Paragon Scientific
N4000
10030 mPa.s (25°C)
101.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16060380
View Documents
Paragon Scientific
S8000
19588 mPa.s (25°C)
92.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16040270
View Documents
Paragon Scientific
N15000
40049 mPa.s (25°C)
87.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16010380
View Documents
Paragon Scientific
S30000
72328 mPa.s (25°C)
87.00 €
500mL
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
Description

Description

We test at Primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy. ASTM D2162 is the only referenced method for the certification of viscosity standards when adopting ASTM test method protocol.

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Density g/mL in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM and IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications

Specifications

500 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • general perpose viscosity standard; UKAS ISO/IEC17025 and ISO guide 34 certified

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : n-hexane

SDS
Documents

Documents

Product Certifications

Nominal values quoted

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.