Learn More
Thermo Scientific™ GeneRuler High Range DNA Ladder
GREENER_CHOICE
Perform fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels.
Brand:  Thermo Scientific™ SM1351
78.39 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 10374880
  • 117.00 € / 50µg
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5hour, 0.4%agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol in the Resources.)
The ladder is supplied with 6X DNA Loading Dye for sample DNA.
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5 hours, 0.4% agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.The ladder is supplied with 6X DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Supplied with loading dye for sample DNA
  • Ideal for fast sizing of high molecular weight DNA fragments
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
GeneRuler
Agarose
High Range DNA Ladder
10171 to 48502 bp
Sustainable packaging
50 μg