Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer
Perform both basic and complex analyses with the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer, an intuitive, reliable solution for education or in the lab.
Brand: Thermo Scientific™ 840-285300
707.75 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Get high precision and functionality in a simple yet sophisticated visible spectrophotometer. The Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer performs both basic and complex analyses for educational or industrial use. Its intuitive design allows students and teachers to focus on scientific data and concepts rather than on learning how to use the instrument. In the lab, technicians of all levels working in routine QA/QC applications can easily obtain credible results to confirm quality or inform next steps. Results can be reported quickly as digital or printed data.
Use with the PV application in the Settings menu of the GENESYS 30 to perform the fundamental PV tests to verify that the instrument is functioning correctly.
Specifications
GENESYS 30 Spectrophotometer Performance Verification Kit |
Product Suggestions
Customers who viewed this item also viewed