Learn More
Thermo Scientific™ GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Brand:  Thermo Scientific™ 840-305700
 View more versions of this product
Product Code. 15896095
  • 4635.00 € / Each
Explore more special offers
Description
Description
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complete 6 mm Fiber Optic Dip Probe system
  • Fiber optic coupler insert for sample compartment
  • 6 mm diameter stainless steel tranflectance probe with replaceable 10 mm pathlength tip
Note: Photometric performance of fiber probe systems is different from cuvette systems and may be impacted by ambient lighting conditions
6 mm
Fiber Optic Dip Probe System
GENESYS 150 and GENESYS 180 Spectrophotometers