Learn More
Genevac™ miVac Centrifugal Concentrators
Choose the ideal concentrator according to workload. Genevac™ miVac Centrifugal Concentrators are modular systems for removing water and solvents from samples. They are available with a range of unique JetRotors™ and vacuum pumps. An optional high-power cold trap is used to condense solvent vapors.
Brand:  Genevac™ QHP-23050-A00
 View more versions of this product
Product Code. 12807633
  • 19434.70 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
miVac Quattro High package
Samples in tubes, microplates and vials
Concentrator with large concentration chamber, high-pressure pump, SpeedTrap, tubing kit
Concentrator: 480W x 594D x 300H mm, Pump: 215W x 394D x 300H mm, SpeedTrap: 212W x 563D x 450H mm
230 V 50/60 Hz 650 w
30°C to 80°C
200 Reaction Vessels (0.5 mL), 20 Microtiter Plates
Max G-force: 250
35 kg (Concentrator), 18 kg (Pump), 25 kg (Speed Trap)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Genevac™ miVac Centrifugal Concentrators > Model: miVac Quattro High kit; Capacity: 200 reaction vessels 0.5mL or 20 microtiter plates; Includes concentrator with large chamber, pump, SpeedTrap, tubing