missing translation for 'onlineSavingsMsg'
Learn More
compliance-icons

Thermo Scientific Chemicals Genipin

Product Code. 17221262
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6902-77-8
Molecular Formula C11H14O5

Specifications

Quantity 250 mg
Melting Point 121.0°C
Formula Weight 226.23
InChI Key AZKVWQKMDGGDSV-BCMRRPTOSA-N
SMILES COC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO
IUPAC Name methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Molecular Weight (g/mol) 226.23
Chemical Name or Material Genipin
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Genipin >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.