missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GenoTube Livestock Swabs

Product Code. 30057769 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 piece
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30057769

Brand: Thermo Scientific™ 9062010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Applied Biosystems™ GenoTube Livestock Swab is easy to use and can be handled by even novice users.

With its proprietary drying system, GenoTube Livestock Swabs rapidly reduce enzyme activity, preventing the breakdown of nucleic acids and proteins that are essential for many laboratory diagnostic procedures. Once dried, collected samples can be stored without refrigeration for simple and cost-effective transportation. In the lab, the samples can be easily reconstituted and used for numerous analyses.

  • Excellent sample quality—fast drying, not influenced by local humidity
  • Convenient versatility—capable of collecting different sample matrices, such as nasal swabs, oral fluids, blood, and fecal samples
  • Cost-effective—low transportation costs because refrigeration or rapid transport not required
  • Individually packaged—allows a unique identifier on both the swab cap and tube for clear identification and also reduces the potential for cross-contamination
  • Superior conservation—no degradation of DNA after long-term storage
  • Sample flexibility—sample can be used for antigen or antibody diagnostics (e.g., PCR, ELISA, etc.)

Genotyping applications

Collecting DNA samples for genotyping and sequencing using the GenoTube Livestock Swab is easy and rapid. After the swab containing the sample is placed back in the tube, the sample is dried, and the DNA in the sample is conserved. The tube can be transported and stored under normal conditions without refrigeration. DNA contained in the sample remains stable for years.

NOTE: this product has a minimum order quantity of 100 pieces.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 GenoTube Livestock Swab

Store at 15–30°C.

Product Line Prionics
Shipping Condition Room Temperature
Quantity 1 piece
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.