missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Gentamicin (10 mg/mL)
Gentamicin (10 mg/mL)
Brand: Gibco™ 15710049
This item is not returnable.
View return policy
Description
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485, cGMP, FDA | |
Gentamicin, 100mL, 10mg/mL, 15 to 30°C Storage Temperature, Antibiotic, Gentamicin Agent, Antibiotic Reagent Type, 24 Months Shelf Life, ISO 13485/cGMP/FDA Certified, Room Temperature/Ambient | |
Liquid | |
Mammalian cell culture, prevention of cell culture contamination |
15 to 30°C | |
10 mg/mL | |
Antibiotic | |
100 mL |
Safety and Handling
Product Identifier
Signal Word
Gefahrenkategorie
Gefahrenhinweis
Sicherheitshinweis
Ergänzende Informationen
- GENTAMICIN (10MG/ML)
Signal Word
- Danger
Gefahrenkategorie
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Gefahrenhinweis
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
- MIXTURE LIST-Contains : gentamicin sulphate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only. Not for use in diagnostic procedures.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur