missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin/Amphotericin Solution

Product Code. 10184583
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10184583

Brand: Gibco™ R01510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gentamicin/Amphotericin Solution

A sterile, concentrated (500x) antibiotic/antimycotic solution of gentamicin and amphotericin B (10 x 1 ml vials/pkg). One Gentamicin/Amphotericin Solution 10-Pak is enough to supplement ten 500 mL bottles of basal medium. Animal origin-free.
TRUSTED_SUSTAINABILITY

Specifications

Type Amphotericin B, Gentamicin
Physical Form Liquid
Sterile Filtered Yes
Concentration 500 X, 125 μg/mL Amphotericin B, 5 mg/mL Gentamicin
Quantity 10 x 1 mL
Validated Application Prevention of Cell Culture Contamination
Product Line Cascade Biologics
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.