Learn More
Gentamicin sulfate, MedChemExpress
MedChemExpress Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.
Brand: Medchem Express HY-A0276
Description
Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.Specifications
Gentamicin sulfate | |
White | |
C(19-21)H(39-43)N5O7.H2SO4 | |
H2O : ≥ 30 mg/mL | |
Research | |
COVID-19-anti-virus |
1405-41-0 | |
Research | |
1 g | |
O=S(O)(O)=O.O[C@]1(C)C(NC)[C@@H](O)[C@@H](O[C@H]2C(N)C[C@H](N)[C@@H](O[C@@H]3[C@H](N)CC[C@@H]([R])O3)[C@@H]2O)OC1.NC[*].C[C@H]([*])N.C[C@H]([*])NC.[R=].[or].[or] | |
Powder |