Learn More
Gentamicin sulfate, MedChemExpress
MedChemExpress Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.
Brand:  Medchem Express HY-A0276
Encompass_Preferred
 View more versions of this product
Product Code. 16471986
  • 79.65 € / 500mg
Explore more special offers
Description
Description
Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.
Specifications
Specifications
Gentamicin sulfate
White
C(19-21)H(39-43)N5O7.H2SO4
H2O : ≥ 30 mg/mL
Research
COVID-19-anti-virus
1405-41-0
Research
500 mg
O=S(O)(O)=O.O[C@]1(C)C(NC)[C@@H](O)[C@@H](O[C@H]2C(N)C[C@H](N)[C@@H](O[C@@H]3[C@H](N)CC[C@@H]([R])O3)[C@@H]2O)OC1.NC[*].C[C@H]([*])N.C[C@H]([*])NC.[R=].[or].[or]
Powder