missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ GeoSampler Soil Sampler

Buerkle™ GeoSampler Soil Sampler is for light soils, meadows and lawns.

Brand:  Buerkle™ 5350-5003

 View more versions of this product

Product Code. 11842303

  • 184.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Soil sampler for light soils, meadows and lawns
  • The foot bar of model 5350-5006 makes work easier
  • Stainless steel
Specifications

Specifications

Geosampler Soil Sampler
No
550 g
56 cm
Stainless Steel
Light Soils, Meadows and Lawns
17 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ GeoSampler Soil Sampler > Length: 56cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.