missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 99%, Thermo Scientific Chemicals

Product Code. 10314702
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 0.8780g/mL
Boiling Point 229.0°C to 230.0°C (757.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
Refractive Index 1.4750 to 1.4790
Quantity 5 g
Beilstein 01,457
Merck Index 15,4437
Specific Gravity 0.878
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and ether, ethanol 50 vol. % (20°C):1:10 v:v
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Geraniol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Geraniol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Geraniol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'