missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geraniol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -15.0°C |
Color | Colorless to Yellow |
Density | 0.8780g/mL |
Boiling Point | 229.0°C to 230.0°C (757.0 mmHg) |
Flash Point | 76°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH |
Refractive Index | 1.4750 to 1.4790 |
Show More |
Safety and Handling
Product Identifier
- Geraniol
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P271-Use only outdoors or in a well-ventilated area.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'