missing translation for 'onlineSavingsMsg'
Learn More

Germanium(II) iodide, 99.99% (metals basis), Thermo Scientific Chemicals

41.00 € - 909.53 €

Chemische Identifikatoren

CAS 13573-08-5
Molecular Formula GeI2
Molecular Weight (g/mol) 326.439
MDL Number MFCD00135537
InChI Key IAGYEMVJHPEPGE-UHFFFAOYSA-N
Synonym germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
PubChem CID 6327215
IUPAC Name diiodogermanium
SMILES [Ge](I)I
Mehr Spezifikationen ansehen

Products 3
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11381539
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014013.77
0.1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11391539
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014013.03
1 g
220.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
11301549
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014013.06
5 g
909.53 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13573-08-5
326.439
IAGYEMVJHPEPGE-UHFFFAOYSA-N
6327215
[Ge](I)I
GeI2
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
diiodogermanium

Specifications

13573-08-5
GeI2
UN3260
IAGYEMVJHPEPGE-UHFFFAOYSA-N
diiodogermanium
6327215
99.99%
5.37g/mL
Germanium(II) iodide
Yellow to Brown
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
[Ge](I)I
326.439
326.4
Air & Moisture Sensitive
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only