missing translation for 'onlineSavingsMsg'
Learn More

Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm, Thermo Scientific Chemicals

Product Code. 11381248
Click to view available options
Quantity:
24 g
120 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Electrical Resistivity: 40 ohm-cm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Color Silver
Physical Form Ingot
Quantity 24 g
Assay Percent Range 99.9999% (Metals basis, 95% N type minimum)
Odor Odorless
Appearance Silver-colored ingot
Chemical Name or Material Germanium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'