missing translation for 'onlineSavingsMsg'
Learn More
Germanium(IV) bromide, 99.999% (metals basis) Product Code.: 15461627

Germanium(IV) bromide, 99.999% (metals basis)

Product Code. 15461627
10 g
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy

Product Code. 15461627

Brand: Thermo Scientific Alfa Aesar 040219.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13450-92-5 | Br4Ge | 392.246 g/mol

It is used as a chemical agent, as a laboratory chemical & for manufacturing of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 187°C
Melting Point 26°C
CAS 13450-92-5
Color White to Gray
Physical Form Low-melting Solid, Liquid
Quantity 10 g
Molecular Formula Br4Ge
MDL Number MFCD00016115
UN Number UN3260
Synonym germanium tetrabromide, germane, tetrabromo, germyl bromide, germanium bromide, germanium iv bromide, germanium, tetrabromo, gebr4, acmc-20aktg, tetrabromine germane
Solubility Information It reacts in water.
InChI Key VJHDVMPJLLGYBL-UHFFFAOYSA-N
SMILES [Ge](Br)(Br)(Br)Br
IUPAC Name tetrabromogermane
Molecular Weight (g/mol) 392.246
PubChem CID 26011
Formula Weight 392.23
Percent Purity 99.999%
Odor Acidic
Sensitivity Moisture sensitive
Density 3.13 g/mL
Assay (metals basis)
Chemical Name or Material Germanium(IV) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium(IV) bromide, 99.999% (metals basis) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.