Learn More
Germanium(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals
323.00 € - 1251.00 €
Chemical Identifiers
CAS | 13450-95-8 |
---|---|
Molecular Formula | GeI4 |
Molecular Weight (g/mol) | 580.248 |
MDL Number | MFCD00016120 |
InChI Key | CUDGTZJYMWAJFV-UHFFFAOYSA-N |
Synonym | germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis |
PubChem CID | 83479 |
IUPAC Name | tetraiodogermane |
SMILES | [Ge](I)(I)(I)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361539
|
Thermo Scientific Alfa Aesar
014009.09 |
10 g |
323.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371539
|
Thermo Scientific Alfa Aesar
014009.18 |
50 g |
1251.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13450-95-8 | |
580.248 | |
CUDGTZJYMWAJFV-UHFFFAOYSA-N | |
83479 | |
[Ge](I)(I)(I)I |
GeI4 | |
MFCD00016120 | |
germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis | |
tetraiodogermane |
Specifications
146°C | |
Crystalline | |
MFCD00016120 | |
germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis | |
CUDGTZJYMWAJFV-UHFFFAOYSA-N | |
tetraiodogermane | |
83479 | |
99.999% | |
4.32 g/mL | |
Germanium(IV) iodide |
13450-95-8 | |
GeI4 | |
UN3260 | |
Soluble in water. | |
[Ge](I)(I)(I)I | |
580.248 | |
580.21 | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 236-613-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only