missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) methoxide, 98%, Thermo Scientific Chemicals

174.87 € - 463.31 €

Chemical Identifiers

CAS 992-91-6
Molecular Formula C4H12GeO4
MDL Number MFCD00014881
Synonym tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488357
SDS View Documents
Thermo Scientific Alfa Aesar
A18437.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11498357
SDS View Documents
Thermo Scientific Alfa Aesar
A18437.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

992-91-6
C4H12GeO4
MFCD00014881
tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv

Specifications

145°C
992-91-6
(CH3O)4Ge
UN2929
tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv
98%
1.4015
1.3254^2^0
-18°C
C4H12GeO4
MFCD00014881
40°C (105°F)
196.73
Like Alcohol
Moisture sensitive
Germanium(IV) methoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H226-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H315-H318-H335

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.