Learn More
GFAP (Glial Fibrillary Acidic Protein) Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
142.00 € - 404.00 €
Specifications
Antigen | GFAP (Glial Fibrillary Acidic Protein) |
---|---|
Clone | ASTRO6 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12649686
|
Epredia
MS-1376-P0 |
Concentrated | 100 μL | N/A | |||||
12679686
|
Epredia
MS-1376-P1ABX |
Concentrated | 100 μL | N/A | |||||
12669686
|
Epredia
MS-1376-PABX |
Concentrated | 200 μL | N/A | |||||
12659686
|
Epredia
MS-1376-P1 |
Concentrated | 500 μL |
283.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12639686
|
Epredia
MS-1376-P |
Concentrated | 1 mL |
404.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12619696
|
Epredia
MS-1376-R7 |
Prediluted | 7 mL |
212.00 €
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. Ab-6 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. Many types of neural tumors such as neuroblastomas, Schwannomas, as well as extra-CNS tumors are not labeled.Host Species: Mouse
Clone: ASTRO6
Isotype: IgG1
Species Reactivity: Human, Pig, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Glial Fibrillary Acidic Protein
Molecular Weight: 51-52 kDa
Positive Control: IMR-5 cells, Brain or astrocytoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
1. Trivino A et al. Vision Res, 1992, 32(9):1601-7.
2. Yachnis AT et al. Journal of Comparative Neurology, 1993, 334(3):356-69.
3. Debus E, et. al. Differentiation, 1983, 25(2):193-203.
Specifications
GFAP (Glial Fibrillary Acidic Protein) | |
Immunofluorescence | |
Unconjugated | |
IVD | |
Chicken, Human, Porcine, Rat | |
IgG1 |
ASTRO6 | |
Monoclonal | |
Mouse | |
Neuroscience | |
Glial Fibrillary Acidic Protein | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.