missing translation for 'onlineSavingsMsg'
Learn More

GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ GFAP (Glial Fibrillary Acidic Protein), Rabbit Polyclonal Antibody.

Brand:  Epredia RB-087-A0

 View more versions of this product

Product Code. 12692777

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Cow, Sheep, Guinea Pig, Chicken, Mouse, Rat and Hamster. Others not known.

Epitope: Not determined

Immunogen: GFAP isolated from cow spinal cord

Molecular Weight: 51-52 kDa

Positive Control: Brain or astrocytoma

Cellular Localization: Cytoplasmic Ab-4 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. In the peripheral nervous system, Schwann cells, satellite cells, and enteric glial cells are stained. Weak staining of axons has been observed. Absorption studies show that this is caused by a cross-reaction with neurofilament protein. Ab-4 is particularly useful for distinguishing neoplasms of astrocytic origin from other neoplasms in the central nervous system by immunohistochemistry.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

GFAP (Glial Fibrillary Acidic Protein)
Polyclonal
Rabbit
Brain or astrocytoma
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
Unconjugated
GFAP isolated from cow spinal cord
100 μL
Neuroscience
Bovine, Chicken, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.